主要文件
Z682292
Ace cold trap outer body only for condenser "CR" assembly
glass, fits Büchi® models R-200/R-205 and R-114/R-144
登录查看公司和协议定价
About This Item
UNSPSC代码:
41121800
NACRES:
NB.43
物料
glass
制造商/商品名称
Büchi equivalent, 11228
正在寻找类似产品? 访问 产品对比指南
一般描述
The cold trap outer body for the Ace condenser "CR" assembly, is glass with a 45/50 inner joint to fit into the distribution head and fits the Büchi® rotary evaporators.
法律信息
Büchi is a registered trademark of Büchi Labortechnik AG
历史批次信息供参考:
分析证书(COA)