主要文件
Z615544
Julabo® bath covers
Lift-up Makrolon cover to fit TW20
登录查看公司和协议定价
About This Item
UNSPSC代码:
41103706
NACRES:
NB.77
包装
pack of 1 ea
制造商/商品名称
Julabo 8970288
长度 × 宽度 × 高度
35 cm × 51 cm × 16 cm
正在寻找类似产品? 访问 产品对比指南
一般描述
Lift-up Makrolon batch cover W 51cm x L35 cm x H 16cm, use to preventnt liquid losses due to evaporation, protects samples from contamination. For use with Julabo eater bath models SW22, SW23.
特点和优势
  • can be used up to +80°C
法律信息
Julabo is a registered trademark of Julabo USA, Inc.
历史批次信息供参考:
分析证书(COA)