主要文件
Z609684
Bohlender screw caps
GL 45 thread
登录查看公司和协议定价
About This Item
UNSPSC代码:
41121800
NACRES:
NB.17
制造商/商品名称
Bohlender
存货情况
not available in USA and Canada
配件
GL 45 thread
正在寻找类似产品? 访问 产品对比指南
一般描述
Made of PPS these screw caps with virgin PTFE membrane gaskets are resistant to high mechanical stresses - even at temperatures upto 250 °C. The gasket consists of PTFE membrane with a multidirectional orientated fibre pattern. Medium will only be exposed to PTFE.
法律信息
Bohlender is a trademark of Bohlender GmbH
历史批次信息供参考:
分析证书(COA)