主要文件
Z566020
Sigma-Aldrich
Pure-Pac® II dispense coupler kit
3/8in. tube end
登录查看公司和协议定价
About This Item
UNSPSC代码:
41100000
NACRES:
NB.01
一般描述
The Sigma-Aldrich Pure-Pac II returnable containers require the Micromatic MacroValve coupler for dispensing chemical. The Dispense Head Coupler Kit includes the MacroValve coupler along with the standard components needed to connect to either 1/2in. or 3/8in. diameter transfer lines. All of the hardware in the kit is supplied in a foam-lined plastic case. See Technical Bulletin AL-236 for assembly instructions.
法律信息
Pure-Pac is a registered trademark of Merck KGaA, Darmstadt, Germany
试剂盒组分也可单独购买
产品编号
说明
化学品安全说明书
  • Z104388PTFE sealing tape, W 1/2 in.化学品安全说明书
  • Male connector with double ferrules and nut, SS, ferrule size 3/8 in., NPTM 1/2 in.
  • Z560723Micromatic MacroValve coupler for returnable containers化学品安全说明书
  • Swagelok® hex reducing coupling, pkg of 1 ea
  • Swagelok® street elbow, pkg of 1 ea
  • Z565547Stainless steel hose connector, 1/4in. male NPT, 1/4in. hose I.D., pkg of 1 ea化学品安全说明书
历史批次信息供参考:
分析证书(COA)