主要文件
Z531367
Sigma-Aldrich
Aldrich® Graham condenser
Overall H 100 mm, joint: ST/NS 14/20
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41121800
NACRES:
NB.43
接合處
joint: ST/NS14/20
存貨情形
not available in EU
總高度
100 mm
正在寻找类似产品? 访问 产品对比指南
相关类别
一般說明
The Graham condenser has a large cooling surface area by allowing coolant to circulate around the inner coil. Condensers have a standard taper outer joint and a lower inner drip-tip joint.
法律資訊
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
历史批次信息供参考:
分析证书(COA)