主要文件
Z510181
Sigma-Aldrich
Large-capacity Kugelrohr glassware sets with System 45 compatible connections
1-L glassware set
登录查看公司和协议定价
About This Item
UNSPSC代码:
41104812
一般描述
Sets contain:
1 – Straight tube with removable in. 1/4 in. I.D. hose connection
1 – Single-bulb ball-tube flask
1 – Round-bottom oven flask
2 – PBT/PTFE screw-thread connectors
3 – Solid-top caps, PBT with PTFE liner
PBT/PTFE connectors resist heat to 200 °C and may be used interchangeably with 1- and 2-liter glassware. Special "reverse threading" on these flasks prevents connections from loosening during rotation.
法律信息
System 45 is a trademark of John Babashak
历史批次信息供参考:
分析证书(COA)