Merck
Z414328
Sigma-Aldrich
Aldrich® septum-inlet adapter with stopcock and side-arm
Plain, joint: ST/NS 24/40, glass stopcock
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41121815
NACRES:
NB.35
材料
glass stopcock
接合處
joint: ST/NS24/40
入口直徑
14 mm
正在寻找类似产品? 访问 产品对比指南
相关类别
一般說明
Requires septum Z564702 or Z564699.
Septum-inlets require septum Z565695 or Z565679 except where noted.
聯結
See the Threaded Ground Glass Tutorial to see how threaded joints work.
法律資訊
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC