主要文件
Z408433
Scienceware® ROTO-FLO flow indicator
fits tubing, 3/16 ‑ 1/4 in., blue wheel
登录查看公司和协议定价
About This Item
UNSPSC代码:
20121904
NACRES:
NB.51
描述
straight tube ends
特点
fits tubing 3/16 ‑ 1/4 in.
制造商/商品名称
Bel-Art
颜色
blue wheel
正在寻找类似产品? 访问 产品对比指南
一般描述
Colored paddle wheel spins freely with liquid or gas flow (range: 0.5 to 10 L/min). Made of clear PS. Not for use with corrosive fluids nor at temperatures above 64 °C.
法律信息
ROTO-FLO is a trademark of SP Scienceware, a division of SP
Scienceware is a registered trademark of SP Scienceware, a division of SP
历史批次信息供参考:
分析证书(COA)