主要文件
Z401870
MSA Defender + faceshield
clear polycarbonate visor
登录查看公司和协议定价
About This Item
UNSPSC代码:
46181802
物料
clear polycarbonate visor
制造商/商品名称
MSA 488132
存货情况
not available in EU
尺寸
8 in. × 16 in. × 0.04 in.
正在寻找类似产品? 访问 产品对比指南
一般描述
  • Lightweight, flexible headbands adjust to accommodate head sizes 61/2 to 8
  • Effective against most flying fragments, particles, splashes, and similar hazards
  • Optional nitrometer chin guard protects against upsweep of particles and splash
  • Use with approved spectacles or goggles
  • Assemblies comply with ANSI Z87.1
Order Defender + headgear assembly, visor and chin guard separately.
历史批次信息供参考:
分析证书(COA)