主要文件
Z334510
Quickfit® three-neck round-bottom flasks with angled necks
center joint: ST/NS 14/23, side joint: ST/NS 14/23
别名:
FR25/1S/11A
登录查看公司和协议定价
About This Item
UNSPSC代码:
41121804
NACRES:
NB.43
物料
borosilicate glass
round bottom flask
three-neck
接合处
center joint: ST/NS14/23
side joint: ST/NS14/23
无菌性
non-sterile
特点
closure type Standard Taper Joint
包装
case of 1
制造商/商品名称
Quickfit FR25/1S/11A
存货情况
not available in USA and Canada
高度
77 mm
正在寻找类似产品? 访问 产品对比指南
一般描述
With three necks fitted with Quickfit, precision-ground sockets. Axes of all three joints meet at bottom of flask.
  • One vertical neck
  • Two angled side necks
法律信息
Quickfit is a registered trademark of SciLabware Limited
历史批次信息供参考:
分析证书(COA)