主要文件
Z301140
Quickfit® expansion adapter
top female joint: ST/NS 29/32, bottom male joint: ST/NS 24/29
别名:
XA43
登录查看公司和协议定价
About This Item
UNSPSC代码:
41121815
NACRES:
NB.35
接合处
female joint: ST/NS top29/32
male joint: ST/NS bottom24/29
制造商/商品名称
Quickfit XA 43
存货情况
not available in USA and Canada
not available in USA and Canada
正在寻找类似产品? 访问 产品对比指南
一般描述
These adapters connect items with larger cone to a piece of glassware with a smaller size socket.
法律信息
Quickfit is a registered trademark of SciLabware Limited
历史批次信息供参考:
分析证书(COA)