主要文件
Z215457
Rigid PTFE sleeve
joint: ST/NS 34/45
别名:
ptfe sleeves
登录查看公司和协议定价
About This Item
UNSPSC代码:
41100000
NACRES:
NB.35
接合处
joint: ST/NS34/45
制造商/商品名称
Cowie
正在寻找类似产品? 访问 产品对比指南
一般描述
Robust, PTFE sleeve with substantial grip-ring for repeated use and ease of handling. Transverse trapezoidal sealing rings provide grease-free, high-vacuum performance at temperatures up to 280°C.
历史批次信息供参考:
分析证书(COA)