主要文件
Z189936
Sigma-Aldrich
Scienceware® pipette support stand
Holds 28 x pipettes
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41122801
NACRES:
NB.01
特點
Holds 28 x pipettes
包裝
pack of 1 ea
製造商/商標名
Bel-Art F18955-0000
直徑 × 高度
7 in. × 7 in.
相关类别
一般說明
  • Scienceware® pipette support stand is made of polypropylene.
  • Ideal for safe and convenient storage of up to 28 pipettes.
  • Features a two-section top plate to hold 10 x 14 mm and 18 x 10 mm pipettes.
  • The 4mm holes in the lower plate protect the tip of the pipettes when orderly placed in the stand.
法律資訊
Scienceware is a registered trademark of SP Scienceware, a division of SP
历史批次信息供参考:
分析证书(COA)