主要文件
Z183431
Sigma-Aldrich
Powerhold clamp
universal
登录查看公司和协议定价
About This Item
UNSPSC代码:
41122413
NACRES:
NB.35
一般描述
With separate safety stop. Used for mounting rods 3/8 to 5/8 in. diameter. Made from high-strength machined aluminum rod with large knobs for gripping power.
历史批次信息供参考:
分析证书(COA)