主要文件
Z137383
Ace Trubore® 3-piece glass stirrer bearing
joint: ST/NS 19/22, fits shaft o.d., 6 mm
登录查看公司和协议定价
About This Item
UNSPSC代码:
41100000
NACRES:
NB.72
接合处
joint: ST/NS19/22
特点
fits shaft o.d. 6 mm
制造商/商品名称
Ace Glass 952910
正在寻找类似产品? 访问 产品对比指南
相关类别
一般描述
Self-aligning, three-piece bearing consisting of a glass inner Trubore bearing, nylon bushing with O-ring, and threaded glass adapter. Inner bearing will spin if shaft binds or will self-align at O-ring if motor is slightly cocked. Inner bearing and bushing can be used with any joint size.
法律信息
Trubore is a registered trademark of Ace Glass, Inc.
历史批次信息供参考:
分析证书(COA)