主要文件
Z131946
Nalgene® calcium chloride drying tube
登录查看公司和协议定价
About This Item
UNSPSC代码:
41100000
包装
pkg of 12 ea
一般描述
PP, single bulb, tubulation at each end. Standard 8 in. length can be reduced to 6 in. or 4 in. by cutting with knife.
法律信息
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
可比产品
产品编号
说明
价格
历史批次信息供参考:
分析证书(COA)