主要文件
TEM-LC300-NI-100
Sigma-Aldrich
Lacey Carbon Supported Nickel Grid
size 300 mesh, hole size 100 μm , Average, box of 25
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41111761
NACRES:
MA.24
包裝
box of 25
孔尺寸
100 μm , Average
尺寸
300 mesh
正在寻找类似产品? 访问 产品对比指南
一般說明
Lacey Carbon Supported Grid: Nickel Grid, 300 mesh. Average hole size is 100 micron. This carbon coated film on a broken pattern consists of woven-mesh-like openings of different sizes and shapes. This type of pattern provides support but does not interfere when observing specimen sections.
历史批次信息供参考:
分析证书(COA)