主要文件
TEM-71166-10
Sigma-Aldrich
Cryo Grid Box
round, w/lid
登录查看公司和协议定价
About This Item
UNSPSC代码:
41111761
NACRES:
MA.24
一般描述
This specimen grid boxe is for storing or transferring cryogenic TEM specimen grids. It features four diamond shaped slots, non-static clear covershield held in place with stainless steel screw, which is tapped in the center of the box. Box fits the FEI Vitrobot, Gatan 626, Gatan 3500, comes with lid, round, this is one unit of cryo grid box.
历史批次信息供参考:
分析证书(COA)