主要文件
SYNF111000
Synthware Dewar flask, tall form with metal housing
flask capacity 1000 mL, tall form with metal housing
别名:
Synthware F111000
登录查看公司和协议定价
About This Item
UNSPSC代码:
41121804
NACRES:
NB.51
物料
aluminum flask
接合处
Joints neck (70)
包装
pkg of 1 ea
制造商/商品名称
Synthware F111000
内径 × 深度
7 cm × 30.2 cm
培养瓶容量
1000 mL
正在寻找类似产品? 访问 产品对比指南
相关类别
一般描述
Dewar flask with weighted metal base for stability and with a full-length aluminum housing which extends to the top rim for added protection.
法律信息
Synthware is a trademark of Kemtech America Inc.
历史批次信息供参考:
分析证书(COA)