主要文件
PL800GN-8
Plas-Labs neoprene glove box gloves
for 1 piece glove set, size 8 (for 8 in. ports)
登录查看公司和协议定价
About This Item
UNSPSC代码:
46181504
NACRES:
NB.35
物料
neoprene
尺寸
8 (for 8 in. ports)
特点
reusable
包装
pack of 1 pair
制造商/商品名称
Plas-Labs 800GN-8
正在寻找类似产品? 访问 产品对比指南
一般描述
Plas-Labs black neoprene rubber gloves for chemical resistance
Note: Order sleeves, gloves, and glove over sleeve connectors separately.
历史批次信息供参考:
分析证书(COA)