主要文件
PL800GB-9-6
Plas-Labs butyl glove box gloves
for 1 piece glove set, size 9 (for 6 in. ports)
登录查看公司和协议定价
About This Item
UNSPSC代码:
46181504
NACRES:
NB.35
物料
butyl rubber
尺寸
9 (for 6 in. ports)
特点
reusable
包装
pack of 1 pair
制造商/商品名称
Plas-Labs 800GB-9-6
正在寻找类似产品? 访问 产品对比指南
一般描述
Plas-Labs black butyl rubber gloves for highest impermeability to water vapor, gases, and toxic fluids
Note: Order sleeves, gloves, and glove over sleeve connectors separately.
历史批次信息供参考:
分析证书(COA)