跳转至内容
Merck
所有图片(1)

主要文件

PH009875

Sigma-Aldrich

(2S)-[(tert-Butoxycarbonyl)amino](4-hydroxyphenyl)ethanoic acid

AldrichCPR

登录查看公司和协议定价


About This Item

线性分子式:
C13H17NO5
CAS号:
UNSPSC代码:
12352106

SMILES字符串

OC([C@@H](NC(OC(C)(C)C)=O)C(C=C1)=CC=C1O)=O

InChI

1S/C13H17NO5/c1-13(2,3)19-12(18)14-10(11(16)17)8-4-6-9(15)7-5-8/h4-7,10,15H,1-3H3,(H,14,18)(H,16,17)/t10-/m0/s1

其他说明

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

历史批次信息供参考:

分析证书(COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our 文件 section.

如需帮助,请联系 客户支持

已有该产品?

在文件库中查找您最近购买产品的文档。

访问文档库

我们的科学家团队拥有各种研究领域经验,包括生命科学、材料科学、化学合成、色谱、分析及许多其他领域.

联系客户支持