主要文件
MT51107873
Mettler Toledo DX series ISE half-cells electrode
Lead selective
登录查看公司和协议定价
About This Item
UNSPSC代码:
39121436
NACRES:
NB.79
物料
membrane (Polymer)
shaft (POM/PVC)
制造商/商品名称
Mettler-Toledo 51107873
测量范围
1.0-1.0-3
温度测量范围
0-50 °C
pH值(酸碱度)
2-8
正在寻找类似产品? 访问 产品对比指南
一般描述
Modular and versatile the METTLER TOLEDO DX series ion-selective half-cells consist of two elements: a universal shaft and an ion-specific membrane module. This module may be exchanged allowing you to measure the ion of your choice. Just order the membrane kit specific for that ion, mount the new module onto the shaft of your ISE half-cell, and you have a new ISE!
electrolyte for reference electrode: 1 mol/L KNO3, ISA solution: 1 mol/L KNO3
历史批次信息供参考:
分析证书(COA)