跳转至内容
Merck

CDS021632

Sigma-Aldrich

1-Benzofuran-3-carboxylic acid

登录查看公司和协议定价


About This Item

经验公式(希尔记法):
C9H6O3
分子量:
162.14
MDL號碼:
分類程式碼代碼:
12352200
PubChem物質ID:

描述

AldrichCPR

SMILES 字串

OC(C1=COC2=C1C=CC=C2)=O

InChI

1S/C9H6O3/c10-9(11)7-5-12-8-4-2-1-3-6(7)8/h1-5H,(H,10,11)

InChI 密鑰

BENJFDPHDCGUAQ-UHFFFAOYSA-N

其他說明

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.
NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY, (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE, OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY, WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

儲存類別代碼

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

水污染物質分類(WGK)

WGK 3

閃點(°F)

Not applicable

閃點(°C)

Not applicable


从最新的版本中选择一种:

分析证书(COA)

Lot/Batch Number

抱歉,我们目前尚未线上提供该产品的COA。

如需帮助,请联系 客户支持

已有该产品?

在文件库中查找您最近购买产品的文档。

访问文档库

我们的科学家团队拥有各种研究领域经验,包括生命科学、材料科学、化学合成、色谱、分析及许多其他领域.

联系技术服务部门