跳转至内容
Merck

CDS012608

Sigma-Aldrich

N,N-dimethylpyrrolidine-2-carboxamide

AldrichCPR

登录查看公司和协议定价


About This Item

经验公式(希尔记法):
C7H14N2O
分子量:
142.20
MDL號碼:
分類程式碼代碼:
12352200
PubChem物質ID:

形狀

solid

SMILES 字串

CN(C)C(=O)C1CCCN1

InChI

1S/C7H14N2O/c1-9(2)7(10)6-4-3-5-8-6/h6,8H,3-5H2,1-2H3

InChI 密鑰

MLLMAIJXIZOSFS-UHFFFAOYSA-N

其他說明

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

象形圖

Exclamation mark

訊號詞

Warning

危險聲明

危險分類

Acute Tox. 4 Oral - Eye Irrit. 2

儲存類別代碼

11 - Combustible Solids

水污染物質分類(WGK)

WGK 3

閃點(°F)

Not applicable

閃點(°C)

Not applicable


历史批次信息供参考:

分析证书(COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our 文件 section.

如需帮助,请联系 客户支持

已有该产品?

在文件库中查找您最近购买产品的文档。

访问文档库

我们的科学家团队拥有各种研究领域经验,包括生命科学、材料科学、化学合成、色谱、分析及许多其他领域.

联系客户支持