主要文件
BR43228
BRAND®密度瓶,Gay-Lussac刻度,未校准
capacity 50 mL, borosilicate glass
别名:
Gay-Lussac
登录查看公司和协议定价
About This Item
UNSPSC代码:
41121800
NACRES:
NB.71
物料
borosilicate glass
glass stopper
narrow-mouth bottle
round bottle (Gay-Lussac)
white bottle
接合处
joint ST/NS neck10/19
特点
with cap
包装
pack of 2 ea
制造商/商品名称
BRAND 43228
容量
50 mL
正在寻找类似产品? 访问 产品对比指南
相关类别
一般描述
DIN ISO 3507, type Gay-Lussac. Stopper NS 10/19 with capillary. Top of stopper ground and polished. Nominal capacity printed on the bottom.
法律信息
BRAND is a registered trademark of BRAND GMBH + CO KG
历史批次信息供参考:
分析证书(COA)
访问文档库
实验方案
Density bottles determine liquid density, calibrated 'to contain' for accuracy, though not volumetric instruments like volumetric flasks.