主要文件
BR144625
BRAND® BISTABIL®磨砂玻璃塞,锥形接头
borosilicate glass 3.3, joint: ST/NS 19/26, (hollow, hexagonal grip)
登录查看公司和协议定价
About This Item
UNSPSC代码:
41121800
NACRES:
NB.43
物料
borosilicate glass 3.3
接合处
joint: ST/NS19/26
制造商/商品名称
BRAND 144625
正在寻找类似产品? 访问 产品对比指南
相关类别
一般描述
Conical ground joint stopper made from Duran®. Hollow with drip tip and hexagonal grip. Meets DIN 12 252.
法律信息
BRAND is a registered trademark of BRAND GMBH + CO KG
Bistabil is a registered trademark of Lenz GmbH
DURAN is a registered trademark of DWK Life Sciences
历史批次信息供参考:
分析证书(COA)