主要文件
BR114106
BRAND®培养管,无唇边
L × W 12 mm × 75 mm, thickness 0.6 mm
别名:
培养管, 试管
登录查看公司和协议定价
About This Item
UNSPSC代码:
41121706
NACRES:
NB.22
物料
round bottom
screw top cap
soda-lime glass tube
straight neck
transparent tube
无菌性
non-sterile
特点
RCF 3,000 × g
neck thread push fit
包装
pack of 250 ea
制造商/商品名称
BRAND 114106
长度 × 宽度
12 mm × 75 mm
容量
12 mL
5.5 mL
厚度
0.6 mm
正在寻找类似产品? 访问 产品对比指南
相关类别
一般描述
Crafted from high-quality glass, these tubes provide excellent clarity and durability, making them ideal for cell culture and other scientific applications. Autoclavable at 121 °C, ensuring easy sterilization.
特点和优势
  • Excellent sample visibility
  • Good chemical resistance
法律信息
BRAND is a registered trademark of BRAND GMBH + CO KG
历史批次信息供参考:
分析证书(COA)