跳转至内容
Merck

BBO000657

Sigma-Aldrich

2-Bromo-6-chloro-4-methylquinoline

登录查看公司和协议定价


About This Item

经验公式(希尔记法):
C10H7BrClN
分子量:
256.53
MDL编号:
UNSPSC代码:
12352200
PubChem化学物质编号:

表单

solid

SMILES字符串

BrC1=NC(C(C(C)=C1)=C2)=CC=C2Cl

InChI

1S/C10H7BrClN/c1-6-4-10(11)13-9-3-2-7(12)5-8(6)9/h2-5H,1H3

InChI key

DRJRVHOBSBPBJS-UHFFFAOYSA-N

其他说明

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

象形图

CorrosionExclamation mark

警示用语:

Danger

危险声明

危险分类

Acute Tox. 4 Oral - Aquatic Chronic 4 - Eye Dam. 1

储存分类代码

11 - Combustible Solids

WGK

WGK 3

闪点(°F)

Not applicable

闪点(°C)

Not applicable


历史批次信息供参考:

分析证书(COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our 文件 section.

如需帮助,请联系 客户支持

已有该产品?

在文件库中查找您最近购买产品的文档。

访问文档库

我们的科学家团队拥有各种研究领域经验,包括生命科学、材料科学、化学合成、色谱、分析及许多其他领域.

联系客户支持