跳转至内容
Merck

ADE001278

Sigma-Aldrich

1-(6-Iodo-2,3-dihydro-1H-pyrido[2,3-b][1,4]oxazin-1-yl)-2,2-dimethylpropan-1-one

AldrichCPR

登录查看公司和协议定价


About This Item

经验公式(希尔记法):
C12H15IN2O2
分子量:
346.16
MDL编号:
UNSPSC代码:
12352200
PubChem化学物质编号:

表单

solid

SMILES字符串

CC(C)(C)C(=O)N1CCOc2nc(I)ccc12

InChI

1S/C12H15IN2O2/c1-12(2,3)11(16)15-6-7-17-10-8(15)4-5-9(13)14-10/h4-5H,6-7H2,1-3H3

InChI key

ZMRLYMRYNIFQRA-UHFFFAOYSA-N

其他说明

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

法律信息

Product of Adesis

储存分类代码

11 - Combustible Solids

WGK

WGK 3

闪点(°F)

Not applicable

闪点(°C)

Not applicable


历史批次信息供参考:

分析证书(COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our 文件 section.

如需帮助,请联系 客户支持

已有该产品?

在文件库中查找您最近购买产品的文档。

访问文档库

我们的科学家团队拥有各种研究领域经验,包括生命科学、材料科学、化学合成、色谱、分析及许多其他领域.

联系客户支持