Skip to Content
Merck
All Photos(2)

Key Documents

MAK195

Sigma-Aldrich

Lipolysis (Adipocyte) Kit

Sufficient for 5 g of tissue

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12352200
NACRES:
NA.25

detection method

colorimetric
fluorometric

relevant disease(s)

endocrinological disorders, diabetes; obesity

storage temp.

−20°C

General description

Lipolysis is the process of hydrolyzing triglycerides to free fatty acids and glycerol. This process involves the action of adipose TG lipase (ATGL), hormone-sensitive lipase (HSL), and monoglyceride lipase. Lipolysis maintains the energy balance during fasting and exercise by providing a substrate for oxidative metabolism. Lipolysis is regulated by nutritional factors and hormones. Problems with the regulation of lipolysis are associated with obesity, diabetes, and metabolic syndromes.

Suitability

This kit is suitable for the isolation of adipocytes from tissue and the measurement of lipolysis.

Principle

The Lipolysis (Adipocyte) Kit provides contains synthetic catecholamine (isoproterenol) that activates β-adrenergic receptors. This results in the activation of adenylate cyclase that converts ATP to cAMP. cAMP then activates the hydrolysis of triglycerides by hormone-sensitive lipase. Lipolysis is determined by measuring a fluorescent (λex = 535/ λem = 587 nm) or colorimetric (570 nm) product proportional to the amount of glycerol present.

Kit Components Only

Product No.
Description

  • Collagenase (0.2%)

  • Collagenase Stop Buffer

  • Adipocyte Wash Buffer

  • Adipocyte Lipolysis Buffer

  • Glycerol Assay Buffer

  • Glycerol Probe, in DMSO

  • Glycerol Enzyme Mix

  • Glycerol Standard, 100 mM

  • Isoproterenol, 10 mM

  • Cell Strainer

See All (10)

replaced by

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

José M Bravo-San Pedro et al.
Cell metabolism, 30(4), 754-767 (2019-08-20)
Autophagy facilitates the adaptation to nutritional stress. Here, we show that short-term starvation of cultured cells or mice caused the autophagy-dependent cellular release of acyl-CoA-binding protein (ACBP, also known as diazepam-binding inhibitor, DBI) and consequent ACBP-mediated feedback inhibition of autophagy.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service