Skip to Content
Merck
All Photos(1)

Key Documents

E2005

Sigma-Aldrich

Eosin 5-isothiocyanate

Synonym(s):

EITC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H7Br4NO5S
CAS Number:
Molecular Weight:
704.96
MDL number:
UNSPSC Code:
12171500
Pricing and availability is not currently available.

storage temp.

2-8°C

SMILES string

OC(=O)c1cc(ccc1C2=C3C=C(Br)C(=O)C(Br)=C3Oc4c(Br)c(O)c(Br)cc24)N=C=S

InChI

1S/C21H7Br4NO5S/c22-12-4-10-14(8-2-1-7(26-6-32)3-9(8)21(29)30)11-5-13(23)18(28)16(25)20(11)31-19(10)15(24)17(12)27/h1-5,27H,(H,29,30)

InChI key

MCXYFYWNOWPDMA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Fluorescent marker useful in studies of triplet state decay and rotational diffusion.

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R J Cherry et al.
Biochemistry, 15(17), 3653-3656 (1976-08-24)
Slow rotational diffusion may be investigated by measuring the decay of dichroism of flash-induced transient absorption changes of suitable probes. The preparation of the covalent "triplets" probe eosin isothiocyanate is described together with investigations of spectroscopic properties of eosin-protein conjugates.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service