Skip to Content
Merck
All Photos(1)

Key Documents

LLCMK2 (Original)

85062804, rhesus monkey kidney, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
biological source:
rhesus monkey kidney
growth mode:
Adherent
karyotype:
2n = 42
morphology:
Epithelial
products:
Not specified
receptors:
Not specified
Pricing and availability is not currently available.

Product Name

LLCMK2 (Original), from rhesus monkey kidney, 85062804, Convention on the International Trade in Endangered Species of Wild Fauna and Flora (CITES)

biological source

rhesus monkey kidney

growth mode

Adherent

karyotype

2n = 42

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Monkey Rhesus kidney

Cell Line Description

Susceptible to a wide range of viruses including enterovirus, rhinovirus, myxovirus and poxvirus groups.

Application

LLCMK2 (Original) has been used to infect and maintain Trypanosoma cruzi parasite culture[1] and as a medium for Sendai virus titer determination.[2] It has also been used for viral culture.[3][4]

Culture Medium

EMEM (EBSS) (M2279) + 2 mM L-Glutamine (G7513) + 1% NEAA (M7145) + 5% FBS / FCS (F2442).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6, i.e., seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37 °C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service