Skip to Content
Merck
All Photos(2)

Documents

AV53846

Sigma-Aldrich

Anti-PRKAA1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-AMPK, Anti-AMPKa1, Anti-MGC33776, Anti-MGC57364, Anti-Protein kinase, AMP-activated, α 1 catalytic subunit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

62 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PRKAA1(5562)

General description

PRKAA1 gene encodes the catalytic α-subunit of 5′ AMP-activated protein kinase (AMPK) that belongs to ser/thr protein kinase family and is localized both in the cytoplasm and in the nucleus. AMPK is a heterotrimeric complex consists of a catalytic α subunit and regulatory β and γ subunits.

Immunogen

Synthetic peptide directed towards the N terminal region of human PRKAA1

Application

Anti-PRKAA1 antibody produced in rabbit is suitable for western blotting at a concentration of 1μg/mL.

Biochem/physiol Actions

AMPK is a cellular energy sensor present in all eukaryotic cells. It is stimulated by high AMP and low ATP concentration and regulates the activities of a number of key metabolic enzymes through allosteric mechanism. Increasing concentrations of AMP during the energy shortage stimulates phosphorylation by an upstream protein kinase and inhibits dephosphorylation, resulting in activation of catabolic pathways and switching off of ATP-consuming biosynthetic pathways. PRKAA1 also facilitates the autophagy-mediated damaged mitochondria clearance for erythrocyte maturation and homeostasis. Additionally, AMPK phosphorylates TSC2 to protect cells from energy deprivation-induced apoptosis and regulates cell growth and survival.

Sequence

Synthetic peptide located within the following region: GELFDYICKNGRKSDVPGVVKTGSTKELDEKESRRLFQQILSGVDYCHRH

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sukriti Krishan et al.
Journal of clinical pathology, 67(9), 758-763 (2014-06-05)
The PRKAA1 gene encodes the catalytic α-subunit of 5′ AMP-activated protein kinase (AMPK). AMPK is a cellular energy sensor that maintains energy homeostasis within the cell and is activated when the AMP/ATP ratio increases. When activated, AMPK increases catabolic processes
Elżbieta Sarnowska et al.
Postepy higieny i medycyny doswiadczalnej (Online), 67, 750-760 (2013-09-11)
AMP-activated protein kinase (AMPK) is one of the major energy sensor at both: cellular and whole body level. It exists as heterotrimer containing three subunits: the catalytic α subunit, β and regulatory γ. AMPK is localized both in the cytoplasm
Ken Inoki et al.
Cell, 115(5), 577-590 (2003-12-04)
Mutations in either the TSC1 or TSC2 tumor suppressor gene are responsible for Tuberous Sclerosis Complex. The gene products of TSC1 and TSC2 form a functional complex and inhibit the phosphorylation of S6K and 4EBP1, two key regulators of translation.
Huaiping Zhu et al.
Autophagy, 10(9), 1522-1534 (2014-07-06)
AMP-activated protein kinase α1 knockout (prkaa1(-/-)) mice manifest splenomegaly and anemia. The underlying molecular mechanisms, however, remain to be established. In this study, we tested the hypothesis that defective autophagy-dependent mitochondrial clearance in prkaa1(-/-) mice exacerbates oxidative stress, thereby enhancing
Abubakar Wani et al.
Autophagy, 17(11), 3813-3832 (2021-01-07)
Alzheimer disease (AD) is usually accompanied by two prominent pathological features, cerebral accumulation of amyloid-β (Aβ) plaques and presence of MAPT/tau neurofibrillary tangles. Dysregulated clearance of Aβ largely contributes to its accumulation and plaque formation in the brain. Macroautophagy/autophagy is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service