Skip to Content
Merck
All Photos(3)

Key Documents

87371

Sigma-Aldrich

Tetrahydrofuran

absolute, over molecular sieve (H2O ≤0.005%), contains ~0.025% 2,6-di-tert-butyl-4-methylphenol as stabilizer, ≥99.5% (GC)

Synonym(s):

Butylene oxide, Oxolane, Tetramethylene oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O
CAS Number:
Molecular Weight:
72.11
Beilstein:
102391
EC Number:
MDL number:
UNSPSC Code:
12191501
PubChem Substance ID:

vapor density

2.5 (vs air)

vapor pressure

114 mmHg ( 15 °C)
143 mmHg ( 20 °C)

grade

absolute

Assay

≥99.5% (GC)

autoignition temp.

610 °F

quality

over molecular sieve (H2O ≤0.005%)

contains

~0.025% 2,6-di-tert-butyl-4-methylphenol as stabilizer

expl. lim.

1.8-11.8 %

impurities

≤0.005% (water)

refractive index

n20/D 1.407 (lit.)

bp

65-67 °C (lit.)

mp

−108 °C (lit.)

density

0.889 g/mL at 25 °C (lit.)

SMILES string

C1CCOC1

InChI

1S/C4H8O/c1-2-4-5-3-1/h1-4H2

InChI key

WYURNTSHIVDZCO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-6.2 °F - closed cup

Flash Point(C)

-21.2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Olav Schiemann et al.
Nature protocols, 2(4), 904-923 (2007-04-21)
In this protocol, we describe the facile synthesis of the nitroxide spin-label 2,2,5,5-tetramethyl-pyrrolin-1-oxyl-3-acetylene (TPA) and then its coupling to DNA/RNA through Sonogashira cross-coupling during automated solid-phase synthesis. Subsequently, we explain how to perform distance measurements between two such spin-labels on
Synthetic routes to tetrahydrofuran, tetrahydropyran, and spiroketal units of polyether antibiotics and a survey of spiroketals of other natural products.
Boivin TLB.
Tetrahedron, 43(15), 3309-3362 (1987)
Eagleson M.
Concise Encyclopedia Chemistry, 152-152 (1994)
Eagleson M.
Concise Encyclopedia Chemistry, 883-883 (1994)
Polyhedral clathrate hydrates. X. Structure of the double hydrate of tetrahydrofuran and hydrogen sulfide.
Mak TCW, et al.
J. Chem. Phys. , 42(8), 2732-2737 (1965)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service