Skip to Content
Merck
All Photos(1)

Key Documents

60103

Sigma-Aldrich

Potassium tert-butoxide solution

purum, ~1 M in THF

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3COK
CAS Number:
Molecular Weight:
112.21
Beilstein:
3556712
MDL number:
UNSPSC Code:
12352000

grade

purum

concentration

~1 M in THF

density

0.910 g/mL at 20 °C

SMILES string

[K+].CC(C)(C)[O-]

InChI

1S/C4H9O.K/c1-4(2,3)5;/h1-3H3;/q-1;+1

InChI key

LPNYRYFBWFDTMA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

nwg


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service