T89400
Tropine hydrate
≥98%
Synonym(s):
2,3-Dihydro-3α-hydroxy-8-methylnortropidine, 3-Tropanol, N-Methyl-8-azabicyclo[3.2.1]octan-3-ol, endo-8-Methyl-8-azabicyclo[3.2.1]octan-3-ol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C8H15NO · xH2O
CAS Number:
Molecular Weight:
141.21 (anhydrous basis)
Beilstein:
80188
EC Number:
MDL number:
UNSPSC Code:
12352103
Assay
≥98%
bp
233 °C (lit.)
mp
50-60 °C (lit.)
density
1.045 g/mL at 25 °C (lit.)
SMILES string
[H]O[H].[H][C@]12CC[C@]([H])(C[C@H](O)C1)N2C
replaced by
Product No.
Description
Pricing
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service