Skip to Content
Merck
All Photos(1)

Key Documents

693340

Sigma-Aldrich

(2S)-3,3-Dimethyl-2-[[(1R,2R)-2-(2-methyl-5-phenyl-1-pyrrolyl)cyclohexyl]thioureido]-N,N-bis(2-isobutyl)butanamide

97%

Synonym(s):

(2S)-3,3-Dimethyl-2-[[[[(1R,2R)-2-(2-methyl-5-phenyl-1H-pyrrol-1-yl)cyclohexyl]amino]thioxomethyl]amino]-N,N-bis(2-methylpropyl)butanamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H50N4OS
CAS Number:
Molecular Weight:
538.83
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

Assay

97%

form

powder

optical activity

[α]22/D -3°, c = 1 in chloroform

mp

143-147 °C

SMILES string

CC(C)CN(CC(C)C)C(=O)[C@@H](NC(=S)N[C@@H]1CCCC[C@H]1n2c(C)ccc2-c3ccccc3)C(C)(C)C

InChI

1S/C32H50N4OS/c1-22(2)20-35(21-23(3)4)30(37)29(32(6,7)8)34-31(38)33-26-16-12-13-17-28(26)36-24(5)18-19-27(36)25-14-10-9-11-15-25/h9-11,14-15,18-19,22-23,26,28-29H,12-13,16-17,20-21H2,1-8H3,(H2,33,34,38)/t26-,28-,29-/m1/s1

InChI key

RIAIKFUBJYQDBD-CRXYYGHGSA-N

Application

Catalyst for acyl-Pictet-Spengler and acyl-Mannich reactions.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mark S Taylor et al.
Journal of the American Chemical Society, 126(34), 10558-10559 (2004-08-26)
The enantioselective cyclization of N-acyliminium ions generated in situ from tryptamine is promoted with high enantioselectivity by a new chiral thiourea catalyst. This represents the first successful system for asymmetric catalysis of the Pictet-Spengler reaction.
Enantioselective thiourea-catalyzed acyl-mannich reactions of isoquinolines.
Mark S Taylor et al.
Angewandte Chemie (International ed. in English), 44(41), 6700-6704 (2005-09-22)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service