Key Documents
CLS2360400
Pyrex® Liebig condenser with bottom ST joint
jacket L 400 mm, bottom joint: ST/NS 24/40
Sinónimos:
Corning® 2360
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
joint
bottom joint: ST/NS24/40
manufacturer/tradename
Corning 2360-400
jacket L
400 mm
General description
tooled neck for No.3 stopper
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.