HSG (HeLa derivative)
95031024, human cervix, Epithelial
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
Recommended Products
product name
HSG (HeLa derivative), 95031024
biological source
human cervix
growth mode
Adherent
karyotype
Not specified
morphology
Epithelial
products
EGF, TGF-beta
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Human cervix carcinoma
Cell Line Description
The cell line HSG was originally thought to be established from a human submandibular gland from a 54 year old Japanese male. The cell line has since been found to be indistinguishable from HeLa by STR PCR DNA profiling. Therefore, the cell line should be considered as derived from HeLa. Ethnicity: Black.
Culture Medium
MEM +2mM Glutamine +1% Non Essential Amino Acids (NEAA) + 10% New Born Calf Serum (NBCS).
Subculture Routine
Split confluent cultures 1:4 to 1:10 ie seeding at 1-3x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2, 37°C
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service