LUDLU-1
92012463, human lung, Epithelial
Synonym(s):
Human LUDLU-1, LUDLU-1 Cell Line, LUDLU-1 DNA
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
eCl@ss:
32190102
Recommended Products
product name
LUDLU-1, 92012463
biological source
human lung
growth mode
Adherent
karyotype
Not specified
morphology
Epithelial
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Human Caucasian lung squamous cell carcinoma
Cell Line Description
Derived from a lung squamous cell carcinoma of a 72 year old Caucasian male. The B-lymphoblastoid cell line AGLCL (Sigma Catalogue number. 89120566) was derived from the same patient. Cells grow as large swollen aggregates, which will detach and eventually grow in suspension.
Application
Control cell line for heterozygosity studies using the AGLCL cell line
Culture Medium
RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).
Subculture Routine
Split sub-confluent cultures (70-80%) 1:5 to 1:10 i.e. seeding at 5x1,000-2x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA, 5% CO2; 37°C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service