S8001U
CpGenome Human Non-Methylated DNA Standard Set
It is intended for use as a negative control in gene methylation studies, such as bisulfite conversion of DNA with the CpGenome Turbo Bisulfite Modification Kit.
Synonym(s):
Human Non-Methylated DNA Standard
About This Item
Recommended Products
Quality Level
species reactivity
human
manufacturer/tradename
CpGenome
Upstate®
application(s)
genomic analysis
shipped in
dry ice
General description
Materials Provided:
One vial containing 5 µg (20 µL) of CpGenome Human Non-Methylated DNA Standard at a concentration of 250 ng/µL.
Validation:
Methylation-specific PCR (MSP) was performed on the CpGenome Human Non-Methylated DNA after bisulfite modification with the CpGenome Turbo Bisulfite kit (Cat. No. S7847). Three sets of primers from the CpG WIZ BRCA1 amplification kit (Cat. No. S7830) were used in this assay: the U primer set, which anneals to unmethylated bisulfite-modified DNA; M primers, which anneal to methylated bisulfite-modified sequences; and W primers, which anneal to unmethylated or methylated DNA that has not undergone bisulfite modification. Only the U primers generated product for this Standard.
CpGenome and CpG WIZ are trademarks of Serologicals Corporation. CpG WIZ Methylation Products apply to technologies exclusively licensed from The Johns Hopkins University School of Medicine. Methylation-specific PCR (MSP) technology is covered under U.S. Patent # 5,786,146.
Legal Information
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service