Z507768
Ace gas dispersion tube with porous fritted glass tip, 15° bend
fits flask, 500 ‑ 2000 mL, porosity 145-174 μm (A)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
feature
fits flask 500 ‑ 2000 mL
manufacturer/tradename
Ace Glass 720403
O.D. × L
7 mm × 250 mm
pore size
145-174 μm porosity (A)
Looking for similar products? Visit Product Comparison Guide
General description
Fit #7 Ace-Thred
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service