Key Documents
Z507695
Ace gas dispersion tube with porous fritted glass tip
Fit #11 Ace-Thred, O.D. × L 10 mm × 150 mm, porosity 10-20 μm (D)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
manufacturer/tradename
Ace Glass 720208
O.D. × L
10 mm × 150 mm
pore size
10-20 μm porosity (D)
Choose from one of the most recent versions:
Certificates of Analysis (COA)