Skip to Content
Merck
All Photos(2)

Key Documents

107999

Sigma-Aldrich

2-Chloroethyl p-toluenesulfonate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4SO3CH2CH2Cl
CAS Number:
Molecular Weight:
234.70
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.529 (lit.)

bp

153 °C/0.3 mmHg (lit.)

density

1.294 g/mL at 25 °C (lit.)

functional group

chloro

storage temp.

2-8°C

SMILES string

Cc1ccc(cc1)S(=O)(=O)OCCCl

InChI

1S/C9H11ClO3S/c1-8-2-4-9(5-3-8)14(11,12)13-7-6-10/h2-5H,6-7H2,1H3

InChI key

ZXNMIUJDTOMBPV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Chloroethyl p-toluenesulfonate was used to develop method for the determination of a variety of hydrophobic aromatic sulfonates by micellar electrokinetic chromatography.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sille Ehala et al.
Journal of chromatography. A, 1161(1-2), 322-326 (2007-07-14)
Two different buffer systems for the separation of 12 aromatic hydrophobic sulfonates by micellar electrokinetic chromatography (MEKC) were developed. The following buffer systems were used: aqueous phosphate buffers containing either cetyltrimethylammonium bromide (CTAB) or sodium dodecyl sulfate (SDS). Eleven aromatic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service