Skip to Content
Merck
All Photos(1)

Key Documents

860736E

Avanti

C24:1 mono-sulfo galactosyl(β) ceramide-d7 (d18:1/24:1)

Avanti Research - A Croda Brand

Synonym(s):

3-O-sulfo-D-galactosyl-ß1-1’-N-nervonoyl-D-erythro-sphingosine-d7 (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C48H87D7N2O11S
Molecular Weight:
914.37
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

ethanol solution

packaging

package of 1 × 100 μg (860736E-100UG)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C (−15°C to −25°C)

Packaging

2 mL Amber Glass Sealed Ampule (860736E-100UG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F

Flash Point(C)

13 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service