Skip to Content
Merck
All Photos(1)

Key Documents

860871P

Avanti

C24:1 Mono-sulfo galactosyl (alpha) ceramide (d18:1/24:1)

Avanti Research - A Croda Brand 860871P, powder

Synonym(s):

3-O-Sulfo-D-galactosyl-α1,1′-N-Nervonoyl-D-erythro-sphingsoine (ammonium salt), (synthetic)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C48H94N2O11S
CAS Number:
Molecular Weight:
907.33
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 1 mg (860871P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860871P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](/C=C/CCCCCCCCCCCCC)(O)[C@@]([H])(NC(CCCCCCCCCCCCC/C=C\CCCCCCCC)=O)CO[C@@H](O1)[C@H](O)[C@@H](OS([O-])(=O)=O)[C@H]([C@H]1CO)O.[NH4+]

InChI

1S/C48H91NO11S.H3N/c1-3-5-7-9-11-13-15-17-18-19-20-21-22-23-24-26-28-30-32-34-36-38-44(52)49-41(42(51)37-35-33-31-29-27-25-16-14-12-10-8-6-4-2)40-58-48-46(54)47(60-61(55,56)57)45(53)43(39-50)59-48;/h17-18,35,37,41-43,45-48,50-51,53-54H,3-16,19-34,36,38-40

InChI key

MQKRPROAXITSNY-BVRPAXLXSA-N

General description

C24:1 Mono-sulfo galactosyl (a) ceramide (d18:1/24:1), also known as 3-O-Sulfo-D-galactosyl-a 1,1′-N-Nervonoyl-D-erythro-sphingsoine, is a synthetic sulfatide that belongs to the class of glycosphingolipids.

Biochem/physiol Actions

Sulfatides play a crucial role in various cellular processes such as protein trafficking, signal transduction and neuronal cell differentiation. Accumulation of 3-O-Sulfogalactosylceramides (sulfatides), due to the deficiency of arylsulfatase A, leads to genetic disease metachromatic leukodystrophy.

Packaging

5 mL Amber Glass Screw Cap Vial (860871P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analysis of complex lipidomes
Uphoff A, et al.
Medical Applications of Mass Spectrometry (2008)
Yanli Cui et al.
Glycoconjugate journal, 25(2), 147-155 (2007-08-19)
3-O-Sulfogalactosylceramides (sulfatides) accumulate in the genetic disease metachromatic leukodystrophy which is due to a defect in the catabolic enzyme, arylsulfatase A. Clinical diagnosis is usually confirmed by in vitro enzymatic deficiency of arylsulfatase A activity. The diagnosis may be complicated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service