Skip to Content
Merck
All Photos(3)

Key Documents

840025P

Avanti

Heart PE

Avanti Research - A Croda Brand 840025P, powder

Synonym(s):

L-α-phosphatidylethanolamine (Heart, Bovine)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 2 × 100 mg (840025P-200mg)
pkg of 1 × 25 mg (840025P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 840025P

lipid type

phosphoglycerides

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC[NH3+])=O)(OC(CCC/C=C\C/C=C\C/C=C\C/C=C\CCCCC)=O)COC(CCCCCCCCCCCCCCCCC)=O

InChI

1S/C43H78NO8P/c1-3-5-7-9-11-13-15-17-19-20-22-24-26-28-30-32-34-36-43(46)52-41(40-51-53(47,48)50-38-37-44)39-49-42(45)35-33-31-29-27-25-23-21-18-16-14-12-10-8-6-4-2/h11,13,17,19,22,24,28,30,41H,3-10,12,14-16,18,20-21,23,25-27,29,31-40,44H2,1-2H3,(H,47,48)

InChI key

ANRKEHNWXKCXDB-BHFWLYLHSA-N

Application

Heart PE has been used in the preparation of liposomes for liposome flotation assays.

Packaging

5 mL Clear Glass Sealed Ampule (840025P-200mg)
5 mL Clear Glass Sealed Ampule (840025P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High-affinity cooperative Ca2+ binding by MICU1-MICU2 serves as an on-off switch for the uniporter
Kamer KJ, et al.
EMBO Reports, 18(8), 1397-1411 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service