Skip to Content
Merck
All Photos(1)

Key Documents

854996

Supelco

Crimp seals with PTFE/silicone septa

silver aluminum seal, open center (10 mm opening), diam. × thickness 20 mm × 3.25 mm, tan PTFE/silicone, pkg of 100 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122001
Pricing and availability is not currently available.

material

silver aluminum seal
tan PTFE/silicone

feature

open center (10 mm opening)
(Agilent HT quality)

hardness

45 (Durometer: shore A)

packaging

pkg of 100 ea

diam. × thickness

20 mm × 3.25 mm

compatibility

for use with 20 mm crimp top vial

Looking for similar products? Visit Product Comparison Guide

Application

Crimp seals with PTFE/silicone septa may be used to make specialized reaction vessel, by fusing them to top autosampler vials, in order to test the accuracy and reproducibility of the tetrabutylammonium fluoride and iodobenzene diacetateTBAF/PhI(OAc)2 method for detecting water in organic solvents.[1] It may also be used to seal various samples, thereby isolating them from atmospheric CO2 for a period of atleast 1 month.[2]

related product

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haoran Sun et al.
Organic letters, 10(20), 4413-4416 (2008-09-13)
The potent dehydrating ability of difluoro(aryl)-lambda (3)-iodanes is exploited to develop a convenient (19)F-NMR-based aquametry method that is more sensitive than coulometric Karl Fischer titration. The key difluoro(aryl)-lambda (3)-iodane reagents are synthesized readily from commercially available and inexpensive precursors.
Rapid and precise d13C measurement of dissolved inorganic carbon in natural waters using liquid chromatography coupled to an isotope-ratio mass spectrometer.
Brandes JA.
Limnology and Oceanography: Methods, 7 (11), 730-739 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service