CLS3206
Corning® Costar® low binding microcentrifuge tubes
0.65mL low binding snap cap microcentrifuge tube, polypropylene, non-sterile, natural, 500/cs
Synonym(s):
centrifuge tubes, microcentrifuge tubes, microfuge tubes
About This Item
Recommended Products
material
cap (Snap Cap)
conical bottom tube
polypropylene
sterility
non-sterile
feature
RCF 17,000 × g
RNase and DNase free
disposable
graduations
packaging
case of 500
pack of 500
manufacturer/tradename
Corning 3206
parameter
17000 × g max. RCF
cap diam.
0.277 in.
tube O.D.
.400 in.
volume
0.65 mL
color
natural
natural
fitting
cap (push fit)
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
These Costar low binding microcentrifuge tubes feature a bonded polymer technology that reduces protein and nucleic acid binding, resulting in better sample recovery. They are made of natural (uncolored) polypropylene and are certified RNase-/DNase-free.
- Supplied nonsterile and are autoclavable
- External graduations and frosted writing spot for easy sample identification
- Positive seal design allows for repeated opening and closing
- Flat cap surface for convenient labeling
- Withstands a maximum RCF of 17,000 x g
Application
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service